ENAH5802E0AE Enamine

N-cyclopropyl-4-fluoro-N'-[1-(propan-2-yl)-1H-pyrazol-5-yl]benzene-1-carboximidamide

  • Cas Number : 1006482-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-14754

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14754
Availability details for
Quantity
  • Availability
  • Add to Cart