ENAH5802E0B4 Enamine

1-aminocyclopent-3-ene-1-carboxylic acid hydrochloride

  • MDL number MFCD28894740
  • Cas Number : 909850-09-5
  • Molecular Weight : 163.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-aminocyclopent-3-ene-1-carboxylic acid hydrochloride
Inchi Code 1S/C6H9NO2.ClH/c7-6(5(8)9)3-1-2-4-6;/h1-2H,3-4,7H2,(H,8,9);1H
Inchi Key VMMUACFKZYTOEE-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7435695

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7435695
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart