ENAH5802E0B7 Enamine

(4E)-2-aminohex-4-enoic acid hydrochloride

  • Molecular Weight : 165.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (E)-2-aminohex-4-enoic acid hydrochloride
Inchi Code 1S/C6H11NO2.ClH/c1-2-3-4-5(7)6(8)9;/h2-3,5H,4,7H2,1H3,(H,8,9);1H/b3-2+;
Inchi Key MTKRKAHKLRCDGI-SQQVDAMQSA-N

Description

Country of Origin UA

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442425
Availability details for
Quantity
  • Availability
  • Add to Cart