ENAH5802E0B8 Enamine

2-[(1r,3r)-3-aminocyclobutyl]acetic acid hydrochloride

  • MDL number MFCD26959151
  • Cas Number : 1523606-40-7
  • Molecular Weight : 165.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-((1r,3r)-3-aminocyclobutyl)acetic acid hydrochloride
Inchi Code 1S/C6H11NO2.ClH/c7-5-1-4(2-5)3-6(8)9;/h4-5H,1-3,7H2,(H,8,9);1H/t4-,5-;
Inchi Key VULUUIGASJOBOK-BPNVWSNHSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7364712

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7364712
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart