ENAH5802E0BA Enamine

3-(aminomethyl)oxetane-3-carboxylic acid

  • MDL number MFCD32197626
  • Cas Number : 1526540-96-4
  • Molecular Weight : 131.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(aminomethyl)oxetane-3-carboxylic acid
Inchi Code 1S/C5H9NO3/c6-1-5(4(7)8)2-9-3-5/h1-3,6H2,(H,7,8)
Inchi Key FMENSXXKJICSDN-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6487727

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6487727
Availability details for
Quantity
  • Availability
  • Add to Cart