ENAH5802E0BF Enamine

lithium(1+) 4-amino-1,3-thiazole-2-carboxylate

  • MDL number MFCD32066674
  • Cas Number : 2361644-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7430013

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430013
Availability details for
Quantity
  • Availability
  • Add to Cart