ENAH5802E0C3 Enamine

(2S)-2-amino-4,4-dimethylpentanoic acid hydrochloride

  • MDL number MFCD30532989
  • Cas Number : 407578-46-5
  • Molecular Weight : 181.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (S)-2-amino-4,4-dimethylpentanoic acid hydrochloride
Inchi Code 1S/C7H15NO2.ClH/c1-7(2,3)4-5(8)6(9)10;/h5H,4,8H2,1-3H3,(H,9,10);1H/t5-;/m0./s1
Inchi Key RCONRVSNXDHMHV-JEDNCBNOSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7437854

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7437854
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart