ENAH5802E0C6 Enamine

2-fluoro-2-(3-hydroxyazetidin-3-yl)acetic acid hydrochloride

  • MDL number MFCD32066170
  • Cas Number : 2361634-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434354
Availability details for
Quantity
  • Availability
  • Add to Cart