ENAH5802E0C7 Enamine

4-(aminomethyl)pyridine-2-carboxylic acid dihydrochloride

  • MDL number MFCD32069111
  • Cas Number : 2375267-93-7
  • Molecular Weight : 225.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(aminomethyl)picolinic acid dihydrochloride

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7438232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438232
Availability details for
Quantity
  • Availability
  • Add to Cart