ENAH5802E0C8 Enamine

5-(aminomethyl)pyridine-2-carboxylic acid dihydrochloride

  • MDL number MFCD31699799
  • Cas Number : 2375260-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723117

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723117
Availability details for
Quantity
  • Availability
  • Add to Cart