ENAH5802E0C9 Enamine

(2R)-2-amino-3-[(S)-methanesulfinyl]propanoic acid

  • MDL number MFCD30582164
  • Cas Number : 32726-14-0
  • Molecular Weight : 151.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2R)-2-amino-3-(methylsulfinyl)propanoic acid
Inchi Code 1S/C4H9NO3S/c1-9(8)2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)/t3-,9?/m0/s1
Inchi Key ZZLHPCSGGOGHFW-BUKSALPDSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7426890

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7426890
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart