ENAH5802E0CB Enamine

2-amino-3,3-difluoropentanoic acid

  • MDL number MFCD31737234
  • Cas Number : 1779684-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1709557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1709557
Availability details for
Quantity
  • Availability
  • Add to Cart