ENAH5802E0CF Enamine

(dimethylphosphoryl)ethene

  • Cas Number : 2180-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6496218

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6496218
Availability details for
Quantity
  • Availability
  • Add to Cart