ENAH5802E0D0 Enamine

6-azaspiro[2.5]octane-5-carboxylic acid hydrochloride

  • MDL number MFCD31744411
  • Cas Number : 2227204-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7441792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7441792
Availability details for
Quantity
  • Availability
  • Add to Cart