ENAH5802E0D2 Enamine

5-azaspiro[3.4]octane-8-carboxylic acid hydrochloride

  • MDL number MFCD32066260
  • Cas Number : 2361645-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434084
Availability details for
Quantity
  • Availability
  • Add to Cart