ENAH5802E0D7 Enamine

4-(aminomethyl)thiophene-2-carboxylic acid hydrochloride

  • MDL number MFCD28894666
  • Cas Number : 2375258-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443289

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443289
Availability details for
Quantity
  • Availability
  • Add to Cart