ENAH5802E0DB Enamine

5,5-dimethylpiperidine-3-carboxylic acid hydrochloride

  • MDL number MFCD32066476
  • Cas Number : 2361635-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7431946

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431946
Availability details for
Quantity
  • Availability
  • Add to Cart