ENAH5802E0DD Enamine

2-(aminomethyl)-1,3-thiazole-5-carboxylic acid hydrochloride

  • Cas Number : 2344678-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7356590

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7356590
Availability details for
Quantity
  • Availability
  • Add to Cart