ENAH5802E0E0 Enamine

2-amino-5-(methylsulfanyl)pentanoic acid hydrochloride

  • Cas Number : 2344677-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7356373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7356373
Availability details for
Quantity
  • Availability
  • Add to Cart