ENAH5802E0E4 Enamine

3-(ethylamino)benzoic acid hydrochloride

  • MDL number MFCD32197778
  • Cas Number : 2375271-47-7
  • Molecular Weight : 201.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(ethylamino)benzoic acid hydrochloride

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7440583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440583
Availability details for
Quantity
  • Availability
  • Add to Cart