ENAH5802E0E6 Enamine

(3R)-3-amino-3-(pyridin-3-yl)propanoic acid dihydrochloride

  • MDL number MFCD03427869
  • Molecular Weight : 239.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3R)-3-amino-3-(3-pyridinyl)propanoic acid dihydrochloride
Inchi Code 1S/C8H10N2O2.2ClH/c9-7(4-8(11)12)6-2-1-3-10-5-6;;/h1-3,5,7H,4,9H2,(H,11,12);2*1H/t7-;;/m1../s1
Inchi Key CTXJKVMJGPHKAI-XCUBXKJBSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7355585

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355585
Availability details for
Quantity
  • Availability
  • Add to Cart