ENAH5802E0EB Enamine

methyl 1-cyano-2-ethenylcyclopropane-1-carboxylate

  • MDL number MFCD27958348
  • Cas Number : 88326-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723006
Availability details for
Quantity
  • Availability
  • Add to Cart