ENAH5802E0EE Enamine

(2S)-2-amino-4-(1H-pyrazol-4-yl)butanoic acid dihydrochloride

  • Cas Number : 2375249-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7442496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442496
Availability details for
Quantity
  • Availability
  • Add to Cart