ENAH5802E0F6 Enamine

2-amino-3-(1-methyl-1H-1,2,3-triazol-4-yl)propanoic acid dihydrochloride

  • Cas Number : 2375267-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443150

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443150
Availability details for
Quantity
  • Availability
  • Add to Cart