ENAH5802E0F8 Enamine

3-amino-4,4,4-trifluoro-3-methylbutanoic acid hydrochloride

  • Cas Number : 856989-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7426233

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426233
Availability details for
Quantity
  • Availability
  • Add to Cart