ENAH5802E0FA Enamine

2-propylpiperidine-2-carboxylic acid hydrochloride

  • Cas Number : 2375274-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7439073

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7439073
Availability details for
Quantity
  • Availability
  • Add to Cart