ENAH5802E100 Enamine

2-methoxy-2-(piperidin-4-yl)acetic acid hydrochloride

  • MDL number MFCD32066212
  • Cas Number : 2361636-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7433634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433634
Availability details for
Quantity
  • Availability
  • Add to Cart