ENAH5802E102 Enamine

5-nitro-3-(prop-2-en-1-yl)-1H-pyrazole

  • MDL number MFCD32198406
  • Cas Number : 2007081-87-8
  • Molecular Weight : 153.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-allyl-5-nitro-1H-pyrazole

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7355037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355037
Availability details for
Quantity
  • Availability
  • Add to Cart