ENAH5802E106 Enamine

2,2-difluoro-2-(piperidin-4-yl)acetic acid hydrochloride

  • MDL number MFCD18433770
  • Cas Number : 1258638-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7429989

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7429989
Availability details for
Quantity
  • Availability
  • Add to Cart