ENAH5802E107 Enamine

2-(1-aminocyclopropyl)pyrimidine-4-carboxylic acid hydrochloride

  • Cas Number : 2344685-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7426009

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426009
Availability details for
Quantity
  • Availability
  • Add to Cart