ENAH5802E109 Enamine

5-(azetidin-3-yl)-1-methyl-1H-pyrazole-4-carboxylic acid; trifluoroacetic acid

  • MDL number MFCD32066889
  • Cas Number : 2361644-39-3
  • Molecular Weight : 295.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2,2-trifluoroacetic acid--5-(azetidin-3-yl)-1-methyl-1H-pyrazole-4-carboxylic acid (1/1)
Inchi Code 1S/C8H11N3O2.C2HF3O2/c1-11-7(5-2-9-3-5)6(4-10-11)8(12)13;3-2(4,5)1(6)7/h4-5,9H,2-3H2,1H3,(H,12,13);(H,6,7)
Inchi Key HYGYKZZQYUCFEO-UHFFFAOYSA-N

Description

Purity 95
Salt Data OC(=O)C(F)(F)F
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7426969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426969
Availability details for
Quantity
  • Availability
  • Add to Cart