ENAH5802E10B Enamine

1-[(azetidin-3-yl)methyl]-1H-1,2,3-triazole-4-carboxylic acid; trifluoroacetic acid

  • MDL number MFCD32069346
  • Cas Number : 2361767-17-9
  • Molecular Weight : 296.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(azetidin-3-ylmethyl)-1H-1,2,3-triazole-4-carboxylic acid--2,2,2-trifluoroacetic acid (1/1)

Description

Purity 95
Salt Data OC(=O)C(F)(F)F
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7435082

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7435082
Availability details for
Quantity
  • Availability
  • Add to Cart