ENAH5802E10D Enamine

5-methyl-4H,5H,6H,7H-[1,2,4]triazolo[1,5-a]pyrimidine-2-carboxylic acid

  • Cas Number : 1702541-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1172668

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1172668
Availability details for
Quantity
  • Availability
  • Add to Cart