ENAH5802E10E Enamine

3-(2-amino-2-carboxyethyl)pyridin-1-ium-1-olate dihydrochloride

  • MDL number MFCD32198003
  • Cas Number : 2375268-80-5
  • Molecular Weight : 255.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-amino-2-carboxyethyl)pyridine 1-oxide dihydrochloride

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7444704

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7444704
Availability details for
Quantity
  • Availability
  • Add to Cart