ENAH5802E10F Enamine

3-(aminomethyl)-5H,6H,7H-pyrrolo[2,1-c][1,2,4]triazole-6-carboxylic acid dihydrochloride

  • MDL number MFCD31924065
  • Cas Number : 2344679-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763992
Availability details for
Quantity
  • Availability
  • Add to Cart