ENAH5802E114 Enamine

decahydroisoquinoline-6-carboxylic acid hydrochloride

  • MDL number MFCD31924712
  • Cas Number : 2344685-64-7
  • Molecular Weight : 219.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6763739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763739
Availability details for
Quantity
  • Availability
  • Add to Cart