ENAH5802E115 Enamine

6-aminospiro[3.5]nonane-6-carboxylic acid hydrochloride

  • MDL number MFCD32066649
  • Cas Number : 2361643-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7433065

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433065
Availability details for
Quantity
  • Availability
  • Add to Cart