ENAH5802E119 Enamine

(2S)-2-amino-4-(trifluoromethoxy)butanoic acid hydrochloride

  • MDL number MFCD32065958

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7356412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7356412
Availability details for
Quantity
  • Availability
  • Add to Cart