ENAH5802E11A Enamine

6-aminonaphthalene-1-carboxylic acid

  • MDL number MFCD11847894
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-amino-1-naphthoic acid
Inchi Code 1S/C11H9NO2/c12-8-4-5-9-7(6-8)2-1-3-10(9)11(13)14/h1-6H,12H2,(H,13,14)
Inchi Key CKSZZOAKPXZMAT-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-307361

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-307361
Availability details for
Quantity
  • Availability
  • Add to Cart