ENAH5802E121 Enamine

2-(aminomethyl)-1H-pyrrolo[2,3-b]pyridine-5-carboxylic acid dihydrochloride

  • MDL number MFCD32068762
  • Cas Number : 2361679-35-6
  • Molecular Weight : 264.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(aminomethyl)-1H-pyrrolo[2,3-b]pyridine-5-carboxylic acid dihydrochloride

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7435324

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7435324
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart