ENAH5802E124 Enamine

2,3,4,5-tetrahydro-1H-1-benzazepine-4-carboxylic acid hydrochloride

  • MDL number MFCD32066211
  • Cas Number : 2361643-58-3
  • Molecular Weight : 227.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7433629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433629
Availability details for
Quantity
  • Availability
  • Add to Cart