ENAH5802E128 Enamine

1-(4-amino-2-fluorophenyl)cyclopropane-1-carboxylic acid hydrochloride

  • Cas Number : 2375270-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443026
Availability details for
Quantity
  • Availability
  • Add to Cart