ENAH5802E12C Enamine

2-(3,3-dimethylbutyl)pyrrolidine-2-carboxylic acid hydrochloride

  • Cas Number : 2375262-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443142

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443142
Availability details for
Quantity
  • Availability
  • Add to Cart