ENAH5802E12F Enamine

ethyl (4E)-2-aminohex-4-enoate hydrochloride

  • Cas Number : 2111873-79-9
  • Molecular Weight : 193.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl (E)-2-aminohex-4-enoate hydrochloride
Inchi Code 1S/C8H15NO2.ClH/c1-3-5-6-7(9)8(10)11-4-2;/h3,5,7H,4,6,9H2,1-2H3;1H/b5-3+;
Inchi Key WGBJYIKGGKQPIJ-WGCWOXMQSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7442842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442842
Availability details for
Quantity
  • Availability
  • Add to Cart