ENAH5802E133 Enamine

6-(prop-1-en-2-yl)-1H-indole

  • MDL number MFCD20688994
  • Cas Number : 865376-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7352630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352630
Availability details for
Quantity
  • Availability
  • Add to Cart