ENAH5802E136 Enamine

methyl 5-fluoro-1,2,3,6-tetrahydropyridine-4-carboxylate hydrochloride

  • MDL number MFCD31924161
  • Cas Number : 2344681-42-9
  • Molecular Weight : 195.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-fluoro-1,2,3,6-tetrahydropyridine-4-carboxylate hydrochloride
Inchi Code 1S/C7H10FNO2.ClH/c1-11-7(10)5-2-3-9-4-6(5)8;/h9H,2-4H2,1H3;1H
Inchi Key INQXCERLPYNGKB-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7425408

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7425408
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart