ENAH5802E137 Enamine

2-(aminomethyl)-3-methyl-1-benzofuran-5-carboxylic acid hydrochloride

  • MDL number MFCD31924583
  • Cas Number : 2344678-71-1
  • Molecular Weight : 241.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(aminomethyl)-3-methylbenzofuran-5-carboxylic acid hydrochloride
Inchi Code 1S/C11H11NO3.ClH/c1-6-8-4-7(11(13)14)2-3-9(8)15-10(6)5-12;/h2-4H,5,12H2,1H3,(H,13,14);1H
Inchi Key YWZPZVDYJODNSB-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7426970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426970
Availability details for
Quantity
  • Availability
  • Add to Cart