ENAH5802E13A Enamine

5-amino-6-(trifluoromethyl)pyridine-2-carboxylic acid

  • MDL number MFCD28727706
  • Cas Number : 1503516-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7426070

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426070
Availability details for
Quantity
  • Availability
  • Add to Cart