ENAH5802E13B Enamine

5-methoxy-2-methyl-2,3-dihydro-1H-indole-3-carboxylic acid hydrochloride

  • MDL number MFCD32069404
  • Cas Number : 2361703-32-2
  • Molecular Weight : 243.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7436544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7436544
Availability details for
Quantity
  • Availability
  • Add to Cart