ENAH5802E13D Enamine

6-methoxy-4-methyl-2,3-dihydro-1H-indole-2-carboxylic acid hydrochloride

  • MDL number MFCD31924200
  • Cas Number : 2344678-47-1
  • Molecular Weight : 243.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methoxy-4-methylindoline-2-carboxylic acid hydrochloride
Inchi Code 1S/C11H13NO3.ClH/c1-6-3-7(15-2)4-9-8(6)5-10(12-9)11(13)14;/h3-4,10,12H,5H2,1-2H3,(H,13,14);1H
Inchi Key IRHAXVWBNJYFJO-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7352577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352577
Availability details for
Quantity
  • Availability
  • Add to Cart